This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02016875_s1
See other IER5L GE Assays ›
Gene Symbol:
Ier5l
Gene Name:
immediate early response 5-like
Gene Aliases:
2610524G09Rik
Chromosome Location:
Chr.2: 30472631 - 30474199 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02016875_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ier5l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030244.3 NP_084520.3 1 1382 169
GenBank mRNA AK012145.1 - 1 1395 169
BC003464.1 - 1 1381 2648900 169
BC010234.1 - 1 1382 3499467 169
BC040084.1 - 1 1382 3499467 169

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ier5l

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103441 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103438 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103454 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103436 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103418 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103452 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103450 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103435 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103453 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103437 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103447 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103439 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103419 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103446 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103442 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103445 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103440 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103449 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121856 BC040084.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121856 AK034055.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121856 BC003464.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121856 BC010234.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121856 NM_030244.3 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113078 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121856 BC040084.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121856 AK034055.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121856 BC003464.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121856 BC010234.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121856 NM_030244.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103457 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103441 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103485 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103488 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103491 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103490 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103453 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103458 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103442 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103476 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103472 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103465 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103467 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103478 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103463 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103430 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103486 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103461 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103450 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103455 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103481 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103484 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103432 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103482 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103459 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103473 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103470 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103462 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103439 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103475 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21103456 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113078 - -
Whole Mouse Genome G4122A A_52_P562641 AK034055.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362514 BC040084.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362514 AK034055.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362514 BC003464.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362514 BC010234.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362514 NM_030244.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110506 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113078 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01604750 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103450 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103452 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103441 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103438 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103440 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103446 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103439 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103453 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103454 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103436 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103447 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103419 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103449 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103445 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103435 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103442 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103437 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21103418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103457 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103478 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103417 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21103378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01771726 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103498 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02177406 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103470 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103433 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103458 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103450 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01604761 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103457 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103479 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103367 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01604749 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103399 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103389 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103503 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103424 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02146065 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103491 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103418 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01771752 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01551693 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103478 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103371 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103369 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103423 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103438 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103500 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103496 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103461 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103390 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103416 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103459 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103449 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103447 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103455 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02194575 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103494 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01771725 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103473 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103439 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103382 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103502 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103402 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103467 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103476 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103440 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01604742 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103400 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103374 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103472 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103376 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103505 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103453 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103441 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01604759 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01179980 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103381 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103410 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103492 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103465 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21103490 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01705201 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10481494 - -
Affymetrix Mouse Gene 1.0 ST Array 10481495 - -
Affymetrix Mouse Gene 1.0 ST Array 10481493 - -
Affymetrix HT MG-430 PM Array Plate 1455881_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1419066_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10481491 - -
Mouse Genome 430 2.0 Array 1419066_at - -
Mouse Genome 430 2.0 Array 1455881_at - -
Mouse Genome 430 2.0 Array 1419066_PM_at - -
Mouse Genome 430 2.0 Array 1455881_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72500_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72500_at - -
Affymetrix Mouse Clariom S Array PSR0200027711.mm.1 NM_030244.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027711.mm.1 NM_030244.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027711.mm.1 TC0200003272.mm.1 MTA TC
Mouse Expression Set 430 1419066_PM_at - -
Mouse Expression Set 430 1419066_at - -
Mouse Expression Set 430 1455881_PM_at - -
Mouse Expression Set 430 1455881_at - -
Murine Genome U74v2 Set 171397_at - -
Murine Genome U74v2 Set 171397_PM_at - -
Mouse Genome 430A 2.0 Array 1419066_at - -
Mouse Genome 430A 2.0 Array 1419066_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419066_at - -
Affymetrix HT Mouse Genome 430A Array 1419066_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1455881_at - -
Affymetrix HT Mouse Genome 430B Array 1455881_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2450154 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002450154 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770609 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2770609 - -
mouse-6 v1.1 (Illumina) scl19470.2_417-S - -
Illumina Sentrix mouse-6.v1 bead array 2450154 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl19470.2_417-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450154 - -
MouseRef-8 v2 Expression BeadChip ILMN_2961005 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2961005 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770609 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19470.2_417-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0