This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01971448_s1
See other A230072C01RIK GE Assays ›
Gene Symbol:
A230072C01Rik
Gene Name:
RIKEN cDNA A230072C01 gene
Gene Aliases:
-
Chromosome Location:
Chr.X: 20961676 - 20987349 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01971448_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A230072C01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027445.1 6 615 154
NR_027446.1 4 861 154
GenBank mRNA AK029359.1 - 6 608 154
AK038883.1 - 4 862 154
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DT931353.1 - 1 92 Chr. X - 20986095 - 20986701 [+] on Build GRCm38 154
GenBankmRNA CK617483.1 - 5 431 Chr. X - 20962075 - 20986302 [+] on Build GRCm38 154

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ubiquitously expressed transcript Uxt 0910002B17Rik -24.0kb Anti-sense Chr. X - 20951665 - 20962035 on NCBI Build GRCm38
Downstream zinc finger protein 182 Zfp182 6330583I20Rik,
Znf182
39.9kb Anti-sense Chr. X - 21026184 - 21062038 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00001545.mm.1 NR_027445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001545.mm.1 NR_027445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001545.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001545.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001545.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001539.mm.1 NR_027445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001539.mm.1 NR_027445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001539.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001543.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001543.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001543.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001542.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001542.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001542.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001544.mm.1 NR_027445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001544.mm.1 NR_027445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001544.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001544.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001544.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001548.mm.1 NR_027445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001548.mm.1 NR_027445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001548.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001548.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001548.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001538.mm.1 NR_027445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001538.mm.1 NR_027445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001538.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001541.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001541.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001541.mm.1 TC0X00000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00001551.mm.1 NR_027445.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001551.mm.1 NR_027445.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001551.mm.1 NR_027446.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001551.mm.1 NR_027446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001551.mm.1 TC0X00000221.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0