Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100040012.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040012.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040008.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040008.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040011.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040011.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040013.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040013.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040016.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040016.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040007.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040007.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040019.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040019.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040024.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040024.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040025.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040025.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040015.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040015.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040031.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040031.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040018.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040018.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040009.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040009.mm.1 |
TC1100004202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040028.mm.1 |
NM_009609.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040028.mm.1 |
TC1100004202.mm.1 |
MTA TC |