This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01961158_s1
See other A130023E24RIK GE Assays ›
Gene Symbol:
A130023E24Rik
Gene Name:
RIKEN cDNA A130023E24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01961158_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A130023E24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK037522.1 - 1 2207 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB155022.2 - 1 618 Chr. 13 - 9129830 - 9130509 [+] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream La ribonucleoprotein domain family,
member 4B
Larp4b A630096F19,
AI256361,
D13Wsu64e,
Larp5
Sense Chr. 13 - 9093814 - 9174406 on NCBI Build GRCm38
Downstream DIP2 disco-interacting protein 2 homolog C (Drosophila) Dip2c 2900024P20Rik,
9630044M06,
mKIAA0934
146.0kb Sense Chr. 13 - 9276503 - 9668928 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Illumina

Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 101230647 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1228715 - -
Illumina Sentrix mouse-6.v1 bead array 101230647 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101230647 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228715 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0