This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01961030_s1
See other D130007C19RIK GE Assays ›
Gene Symbol:
D130007C19Rik
Gene Name:
RIKEN cDNA D130007C19 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 123902072 - 123904860 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01961030_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130007C19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK051152.1 - 1 939 120
BC147505.1 - 1 342 120

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23265181 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23265180 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23265182 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156119 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265182 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265167 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265161 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265147 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265175 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265165 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265151 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265170 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265177 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265178 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265179 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265154 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265162 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265173 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265149 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265181 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265172 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265156 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23265176 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156119 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156119 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23265181 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23265182 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23265180 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100940286 - -
Illumina mouse-6 v1.0 expression beadchip 100940286 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1238543 - -
Illumina Sentrix mouse-6.v1 bead array 100940286 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100940286 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238543 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0