This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01731211_gH
See other UBE2DNL2 GE Assays ›
Gene Symbol:
Ube2dnl2
Gene Name:
ubiquitin-conjugating enzyme E2D N-terminal like 2
Gene Aliases:
4930524E20Rik
Chromosome Location:
Chr.X: 114907582 - 114908510 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01731211_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2dnl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081661.1 NP_001075130.1 1 - 2 193 131
GenBank mRNA AK015883.1 - 1 - 2 194 131

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001081661.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437683 NM_001081661.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437683 AK015883.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437683 NM_001081661.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437683 AK015883.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874877 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437683 NM_001081661.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437683 AK015883.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437683 NM_001081661.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437683 AK015883.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P437683 NM_001081661.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P437683 AK015883.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P437683 NM_001081661.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P437683 AK015883.1 mouse_transcriptome
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611565 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611541 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611530 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32611535 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611521 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611536 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611512 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611554 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611506 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611578 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611556 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611542 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611509 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611567 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611560 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611539 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611553 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611523 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611583 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611581 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611515 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611577 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611517 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611574 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611524 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611530 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611573 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611571 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611586 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611585 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14837370 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611518 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15059896 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611543 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15198745 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611572 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611519 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611508 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611570 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611547 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611582 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611565 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32611533 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14967476 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10601536 - -
Affymetrix Mouse Gene 1.0 ST Array 10601535 - -
Affymetrix Mouse Gene 1.1 ST Array 10601534 - -
Affymetrix Mouse Clariom S Array PSR0X00007469.mm.1 NM_001081661.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007469.mm.1 NM_001081661.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007469.mm.1 TC0X00001147.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00007470.mm.1 NM_001081661.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007470.mm.1 NM_001081661.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007470.mm.1 TC0X00001147.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 498187 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_1215307 - -
MouseRef-8 v2 Expression BeadChip ILMN_3009928 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009928 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0