This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01721874_m1
See other PIR GE Assays ›
Gene Symbol:
Pir
Gene Name:
pirin
Gene Aliases:
2310042L19Rik, Pirnl
Chromosome Location:
Chr.X: 164269431 - 164373013 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01721874_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pir
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301402.1 NP_001288331.1 3 - 4 423 96
NM_027153.3 NP_081429.1 4 - 5 552 96
GenBank mRNA AK009757.1 - 4 - 5 495 96
AK082922.1 - 4 - 5 437 96
AK090023.1 - 4 - 5 409 96
AK138314.1 - 4 - 5 548 96
AK151301.1 - 4 - 5 455 96
BC024062.1 - 4 - 5 458 5067536 96
BY720928.1 - 4 - 5 553 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001665.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pir

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
myeloid cell differentiation
monocyte differentiation
oxidation-reduction process
regulation of nucleic acid-templated transcription
transcription cofactor activity
quercetin 2,3-dioxygenase activity
oxidoreductase activity
metal ion binding
dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00010651.mm.1 NM_027153.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010651.mm.1 NM_027153.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010651.mm.1 TC0X00001665.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0