This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01721423_m1
See other 5430401H09RIK GE Assays ›
Gene Symbol:
5430401H09Rik
Gene Name:
RIKEN cDNA 5430401H09 gene
Gene Aliases:
AW987716
Chromosome Location:
Chr.12: 32096593 - 32124760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01721423_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5430401H09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK028422.1 - 1 - 2 83 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX524907.1 - 1 - 2 47 Chr. 12 - 32097257 - 32123667 [-] on Build GRCm38 73
GenBankmRNA AW989880.1 - 1 - 2 46 Chr. 12 - 32097349 - 32123666 [-] on Build GRCm38 73
GenBankmRNA BB570574.2 - 1 - 2 83 Chr. 12 - 32115334 - 32123702 [-] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein kinase,
cAMP dependent regulatory,
type II beta
Prkar2b AI451071,
AW061005,
PKARIIbeta,
Pkarb2,
RII(beta)
-54.0kb Sense Chr. 12 - 31958450 - 32061343 on NCBI Build GRCm38
Downstream phosphoinositide-3-kinase,
catalytic,
gamma polypeptide
Pik3cg 5830428L06Rik,
PI3Kgamma,
p110gamma
49.4kb Sense Chr. 12 - 32173129 - 32208677 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2357303 AW987716.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2357303 AK017255.1 mouse_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0