This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01700207_m1
See other GJB1 GE Assays ›
Gene Symbol:
Gjb1
Gene Name:
gap junction protein, beta 1
Gene Aliases:
Cnx32, Cx32
Chromosome Location:
Chr.X: 101376376 - 101385629 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01700207_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC026833.1 - 1 - 2 28 3483365 59
BY087537.1 - 1 - 2 59 59
BY105229.1 - 1 - 2 93 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC026833.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

gap junction

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006445.mm.1 NM_008124.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006445.mm.1 TC0X00000973.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 810388 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0