This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01611746_g1
See other MRPS26 GE Assays ›
Gene Symbol:
Mrps26
Gene Name:
mitochondrial ribosomal protein S26
Gene Aliases:
AI648866, GI008, MRP-S13, MRP-S26, Rpms13
Chromosome Location:
Chr.2: 130563757 - 130565394 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01611746_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrps26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207207.1 NP_997090.1 1 - 2 236 107
GenBank mRNA AK131626.1 - 1 - 2 246 107
AK169837.1 - 1 - 2 249 107
BC048517.1 - 1 - 2 236 6706680 107

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrps26

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630447 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630445 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 NM_207207.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 AK131626.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 AK169837.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 AK213019.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 BC048517.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 NM_207207.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 AK131626.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 AK169837.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 AK213019.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339713 BC048517.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992989 AK131626.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140802 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992989 AK131626.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 NM_207207.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 AK131626.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 AK169837.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 AK213019.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 BC048517.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 NM_207207.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 AK131626.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 AK169837.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 AK213019.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339713 BC048517.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 NM_207207.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 AK131626.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 AK169837.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 AK213019.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 BC048517.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 NM_207207.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 AK131626.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 AK169837.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 AK213019.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339713 BC048517.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630426 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630431 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630451 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630422 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630423 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630454 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630430 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630421 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630424 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630442 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630434 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630452 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630428 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630437 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630460 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630433 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140802 - -
Whole Mouse Genome G4122A A_51_P339713 NM_207207.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 AK131626.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 AK169837.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 AK213019.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 BC048517.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 NM_207207.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 AK131626.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 AK169837.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 AK213019.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339713 BC048517.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P542958 NM_207207.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P542958 AK131626.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P542958 AK169837.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P542958 AK213019.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P542958 BC048517.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112542 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137700 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140802 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01896236 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01896252 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01896251 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01896234 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630445 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630499 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630422 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896236 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01228898 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896258 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01279555 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01279558 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896252 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630452 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630454 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630442 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630411 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896231 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630428 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630426 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01528593 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02164985 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630414 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01992608 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896248 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896235 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01536740 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02063828 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01286708 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896233 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01279557 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01536741 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01992606 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01992610 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01896251 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01279553 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01286709 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01992611 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630408 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21630412 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10476190 - -
Affymetrix Mouse Gene 1.0 ST Array 10476191 - -
Affymetrix HT MG-430 PM Array Plate 1433878_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10476189 - -
Mouse Genome 430 2.0 Array 1433878_PM_at - -
Mouse Genome 430 2.0 Array 1433878_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 99045_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 99045_at - -
Affymetrix Mouse Clariom S Array PSR0200015796.mm.1 NM_207207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015796.mm.1 NM_207207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015796.mm.1 TC0200001990.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200015793.mm.1 NM_207207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015793.mm.1 NM_207207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015793.mm.1 TC0200001990.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200015797.mm.1 NM_207207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015797.mm.1 NM_207207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015797.mm.1 TC0200001990.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200015794.mm.1 NM_207207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015794.mm.1 NM_207207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015794.mm.1 TC0200001990.mm.1 MTA TC
Affymetrix HT Mouse Genome 430A Array 1433878_at - -
Affymetrix HT Mouse Genome 430A Array 1433878_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 895706 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4120113 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004120113 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2767118 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2767118 - -
mouse-6 v1.1 (Illumina) scl099045.1_16-S - -
Illumina Sentrix mouse-6.v1 bead array 4120113 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl099045.1_16-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120113 - -
MouseRef-8 v2 Expression BeadChip ILMN_3005827 - -
MouseRef-8 v2 Expression BeadChip ILMN_3005828 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3005827 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3005828 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl099045.1_16-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0