This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01545547_m1
See other TMEM161B GE Assays ›
Gene Symbol:
Tmem161b
Gene Name:
transmembrane protein 161B
Gene Aliases:
2810446P07Rik, AI843389
Chromosome Location:
Chr.13: 84222187 - 84295966 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01545547_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem161b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029418.1 1 - 2 136 85
GenBank mRNA AK013305.1 - 1 - 2 107 85
AK051974.1 - 1 - 2 107 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK013305.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 NM_175187.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 NM_175187.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 NR_029418.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 NR_029418.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 NR_029419.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 NR_029419.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006338.mm.1 TC1300000983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 NM_175187.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 NM_175187.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 NR_029418.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 NR_029418.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 NR_029419.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 NR_029419.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006340.mm.1 TC1300000983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 NM_175187.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 NM_175187.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 NR_029418.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 NR_029418.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 NR_029419.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 NR_029419.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006341.mm.1 TC1300000983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 NM_175187.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 NM_175187.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 NR_029418.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 NR_029418.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 NR_029419.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 NR_029419.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006337.mm.1 TC1300000983.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0