This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01352686_g1
See other PQBP1 GE Assays ›
Gene Symbol:
Pqbp1
Gene Name:
polyglutamine binding protein 1
Gene Aliases:
PQBP-1, Sfc2, npw38
Chromosome Location:
Chr.X: 7894519 - 7899269 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01352686_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pqbp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252529.1 NP_001239458.1 1 - 2 102 72
GenBank mRNA AK077652.1 - 1 - 2 61 72
BC051673.1 - 1 - 2 41 30015428 72
BQ550428.1 - 1 - 2 107 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK077652.1, BC051673.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

alternative mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
neuron projection development
stress granule assembly
regulation of RNA splicing
regulation of dendrite morphogenesis
cilium morphogenesis
protein C-terminus binding
ribonucleoprotein complex binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00011313.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011313.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011313.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011301.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011311.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011311.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011311.mm.1 TC0X00001784.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 910025 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0