This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01352023_m1
See other MYRIP GE Assays ›
Gene Symbol:
Myrip
Gene Name:
myosin VIIA and Rab interacting protein
Gene Aliases:
A230081N12Rik, Slac2-c, Slac2c
Chromosome Location:
Chr.9: 120303919 - 120476101 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01352023_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Myrip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144557.5 NP_653140.1 11 - 12 2053 101
GenBank mRNA AB083782.1 - 10 - 11 1892 101
AF396688.1 - 10 - 11 1915 101
AK038988.1 - 11 - 12 2007 101
AK083225.1 - 11 - 12 2054 101
AK148358.1 - 11 - 12 2170 101
AY099470.1 - 10 - 11 1928 101
BC129851.1 - 10 - 11 1937 40109680 101
BC129852.1 - 10 - 11 1937 40109682 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001600.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Myrip

Panther Classification:

Molecular Function -

myelin protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle transport along actin filament
positive regulation of insulin secretion
actin binding
protein binding
myosin binding
Rab GTPase binding
metal ion binding
protein kinase A binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900013366.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013366.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013366.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013369.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013369.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013369.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013367.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013367.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013367.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013376.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013376.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013376.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013364.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013364.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013364.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013371.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013371.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013371.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013375.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013375.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013375.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013378.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013378.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013378.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013370.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013370.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013370.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013362.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013362.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013362.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013372.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013372.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013372.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013377.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013377.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013377.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013361.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013361.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013361.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013368.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013368.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013368.mm.1 TC0900001600.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900013374.mm.1 NM_144557.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900013374.mm.1 NM_144557.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900013374.mm.1 TC0900001600.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 757355 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0