This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351271_m1
See other HSDL2 GE Assays ›
Gene Symbol:
Hsdl2
Gene Name:
hydroxysteroid dehydrogenase like 2
Gene Aliases:
2610207I16Rik
Chromosome Location:
Chr.4: 59580458 - 59618694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351271_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsdl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301367.1 NP_001288296.1 3 - 4 478 105
NM_024255.3 NP_077217.2 2 - 3 380 105
GenBank mRNA AJ293845.1 - 3 - 4 401 105
AK011910.1 - 2 - 3 338 105
AK085899.1 - 2 - 3 349 105
AK136773.1 - 2 - 3 347 105
AK144948.1 - 2 - 3 296 105
AK145230.1 - 2 - 3 240 105
AK202946.1 - 1 - 2 95 105
AK205169.1 - 1 - 2 95 105
AK211849.1 - 1 - 2 95 105
AK213102.1 - 1 - 2 87 105
AK215396.1 - 1 - 2 95 105
AY725196.1 - 2 - 3 237 105
BC002211.1 - 2 - 3 253 3490303 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AJ293845.1, NM_001301367.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsdl2

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
oxidation-reduction process
molecular_function
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400004662.mm.1 NM_024255.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400004662.mm.1 NM_024255.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400004662.mm.1 TC0400000659.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0