This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351210_m1
See other SLC6A19 GE Assays ›
Gene Symbol:
Slc6a19
Gene Name:
solute carrier family 6 (neurotransmitter transporter), member 19
Gene Aliases:
4632401C08Rik, B<0>AT1
Chromosome Location:
Chr.13: 73679745 - 73700695 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351210_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc6a19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028878.3 NP_083154.1 8 - 9 1232 107
GenBank mRNA AJ633679.1 - 8 - 9 1241 107
AK014544.1 - 8 - 9 1233 107
AK028784.1 - 8 - 9 1233 107
AK128923.1 - 7 - 8 1575 107
AK162399.1 - 7 - 8 977 107
BC107411.1 - 8 - 9 1233 40059042 107
BC107412.2 - 8 - 9 1237 40059050 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002303.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc6a19

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
neurotransmitter transport
amino acid transport
neutral amino acid transport
neurotransmitter:sodium symporter activity
protein binding
neutral amino acid transmembrane transporter activity
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300015099.mm.1 NM_028878.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300015099.mm.1 NM_028878.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300015099.mm.1 TC1300002303.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300015098.mm.1 NM_028878.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300015098.mm.1 NM_028878.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300015098.mm.1 TC1300002303.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0