This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350814_m1
See other SLC44A1 GE Assays ›
Gene Symbol:
Slc44a1
Gene Name:
solute carrier family 44, member 1
Gene Aliases:
2210409B22Rik, 4833416H08Rik, AW547365, CHTL1, CTL1, Cdw92
Chromosome Location:
Chr.4: 53440413 - 53622478 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350814_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc44a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159633.1 NP_001153105.1 4 - 5 669 100
NM_133891.3 NP_598652.3 4 - 5 669 100
GenBank mRNA AY249865.1 - 4 - 5 425 100
AY249866.1 - 4 - 5 398 100
BC113167.1 - 4 - 5 425 40059854 100
BC113169.1 - 4 - 5 425 40059860 100
CF532681.1 - 3 - 4 300 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000580.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc44a1

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
organic cation transport
choline transport
ammonium transmembrane transport
choline transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400004221.mm.1 NM_001159633.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400004221.mm.1 NM_001159633.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400004221.mm.1 NM_133891.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400004221.mm.1 NM_133891.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400004221.mm.1 TC0400000580.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0