This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350070_m1
See other PIGZ GE Assays ›
Gene Symbol:
Pigz
Gene Name:
phosphatidylinositol glycan anchor biosynthesis, class Z
Gene Aliases:
F630022B06Rik
Chromosome Location:
Chr.16: 31926073 - 31946048 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350070_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pigz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172822.3 NP_766410.2 2 - 3 392 81
GenBank mRNA AK087516.1 - 2 - 3 275 81
AK089183.1 - 2 - 3 291 81
BC117748.2 - 1 - 2 212 40090477 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pigz

Panther Classification:

Molecular Function -

glycosyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor biosynthetic process
mannosylation
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0