This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01349760_m1
See other GOLGB1 GE Assays ›
Gene Symbol:
Golgb1
Gene Name:
golgi autoantigen, golgin subfamily b, macrogolgin 1
Gene Aliases:
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151
Chromosome Location:
Chr.16: 36875173 - 36933085 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01349760_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Golgb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030035.1 NP_084311.1 15 - 16 8977 66
GenBank mRNA AK220188.1 - 3 - 4 4832 66
BC040461.1 - 3 - 4 2297 66
BC042759.1 - 2 - 3 312 66
BC060254.1 - 2 - 3 1401 66
BC145221.1 - 13 - 14 1919 9053772 66
BC150731.1 - 15 - 16 8954 9087642 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0