This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01347922_m1
See other SLC4A4 GE Assays ›
Gene Symbol:
Slc4a4
Gene Name:
solute carrier family 4 (anion exchanger), member 4
Gene Aliases:
AI835705, NBC, NBC1
Chromosome Location:
Chr.5: 88887247 - 89239656 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01347922_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136260.1 NP_001129732.1 11 - 12 1389 128
NM_001197147.1 NP_001184076.1 11 - 12 1362 128
NM_018760.2 NP_061230.2 11 - 12 1389 128
GenBank mRNA AF141934.3 - 8 - 9 1191 128
BC026592.1 - 2 - 3 182 128
BC129863.1 - 11 - 12 1351 128
BC129864.1 - 11 - 12 1350 128
BC148292.2 - 11 - 12 1325 40110074 128
HQ018820.1 - 8 - 9 1226 128
HQ204220.1 - 11 - 12 1333 128
HQ285250.1 - 8 - 9 1202 128

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a4

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
anion transport
regulation of pH
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
anion transmembrane transport
transporter activity
inorganic anion exchanger activity
protein binding
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0