This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343840_m1
See other C330027C09RIK GE Assays ›
Gene Symbol:
C330027C09Rik
Gene Name:
RIKEN cDNA C330027C09 gene
Gene Aliases:
AA408511, AU018569, Cip2a, Kiaa1524
Chromosome Location:
Chr.16: 48994188 - 49019711 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343840_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330027C09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172616.2 NP_766204.2 2 - 3 339 131
GenBank mRNA AK049356.1 - 2 - 3 341 131
AK160639.1 - 2 - 3 325 131
AK173190.1 - 1 - 2 77 131
BC137719.1 - 2 - 3 348 8860738 131

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000700.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene C330027C09Rik

Gene Ontology Categories:

Function(s)

spermatogenesis
positive regulation of neural precursor cell proliferation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600006102.mm.1 NM_172616.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600006102.mm.1 NM_172616.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600006102.mm.1 TC1600000700.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0