This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343784_m1
See other SIDT1 GE Assays ›
Gene Symbol:
Sidt1
Gene Name:
SID1 transmembrane family, member 1
Gene Aliases:
AW045928, B830021E24Rik
Chromosome Location:
Chr.16: 44240177 - 44333328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343784_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sidt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159419.1 NP_001152891.1 2 - 3 651 87
NM_198034.3 NP_932151.2 2 - 3 651 87
GenBank mRNA AK046836.1 - 2 - 3 87
AK171920.1 - 2 - 3 804 87
BC079584.1 - 2 - 3 651 5704672 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001642.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sidt1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
dsRNA transport
molecular_function
RNA transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600013413.mm.1 NM_001159419.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013413.mm.1 NM_001159419.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013413.mm.1 NM_198034.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013413.mm.1 NM_198034.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013413.mm.1 TC1600001642.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0