This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343454_m1
See other SACM1L GE Assays ›
Gene Symbol:
Sacm1l
Gene Name:
SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae)
Gene Aliases:
SAC1, Sac1p, mKIAA0851
Chromosome Location:
Chr.9: 123529882 - 123592598 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343454_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sacm1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030692.4 NP_109617.1 16 - 17 1391 71
GenBank mRNA AJ245720.1 - 16 - 17 1521 71
AJ297526.1 - 16 - 17 1392 71
AK089174.1 - 16 - 17 1475 71
AK122390.1 - 16 - 17 1456 71
AK154759.1 - 16 - 17 1491 71
AK161111.1 - 16 - 17 1425 71
AK171080.1 - 16 - 17 1475 71
BC005489.1 - 7 - 8 600 71
BC117752.1 - 16 - 17 1392 40090556 71
BC117753.1 - 16 - 17 1392 40090558 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001666.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sacm1l

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol dephosphorylation
phosphatidylinositol-3-phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
phosphohistidine phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
phosphatidylinositol bisphosphate phosphatase activity
phosphoric ester hydrolase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
IDP phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900014021.mm.1 NM_030692.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900014021.mm.1 NM_030692.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900014021.mm.1 TC0900001666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900014020.mm.1 NM_030692.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900014020.mm.1 NM_030692.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900014020.mm.1 TC0900001666.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0