This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343354_m1
See other ADCY5 GE Assays ›
Gene Symbol:
Adcy5
Gene Name:
adenylate cyclase 5
Gene Aliases:
AW121902, Ac5
Chromosome Location:
Chr.16: 35155636 - 35304549 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343354_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adcy5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012765.4 NP_001012783.3 8 - 9 2559 68
GenBank mRNA AK147590.1 - 2662 68
AK147649.1 - 3319 68
AK160942.1 - 8 - 9 2559 68
BC035550.1 - 697 68
BC090846.1 - 8 - 9 2294 30363147 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adcy5

Gene Ontology Categories:

Function(s) Process(es)

adenosine receptor signaling pathway
cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
adenylate cyclase-inhibiting dopamine receptor signaling pathway
locomotory behavior
cyclic nucleotide biosynthetic process
intracellular signal transduction
neuromuscular process controlling balance
nucleotide binding
adenylate cyclase activity
protein binding
ATP binding
adenylate cyclase binding
calcium- and calmodulin-responsive adenylate cyclase activity
lyase activity
phosphorus-oxygen lyase activity
metal ion binding
protein heterodimerization activity
scaffold protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0