This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341526_g1
See other SLC25A51 GE Assays ›
Gene Symbol:
Slc25a51
Gene Name:
solute carrier family 25, member 51
Gene Aliases:
9130208E07Rik, D130005A03Rik, Gm138, Mcart1
Chromosome Location:
Chr.4: 45395923 - 45408783 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341526_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009949.3 NP_001009949.3 1 - 2 75 78
GenBank mRNA AK051139.1 - 71 78
AK146788.1 - 1 - 2 37 78
AK150362.1 - 1 - 2 33 78
AK151688.1 - 1 - 2 33 78
AK152171.1 - 1 - 2 33 78
AK156168.1 - 1 - 2 33 78
AK156342.1 - 1 - 2 33 78
AK164726.1 - 1 - 2 56 78
AK165253.1 - 1 - 2 71 78

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK146788.1,AK150362.1,AK151688.1,AK152171.1,AK156168.1,AK165253.1,NM_001009949.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a51

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400021029.mm.1 NM_001009949.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021029.mm.1 NM_001009949.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021029.mm.1 TC0400002578.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021022.mm.1 NM_001009949.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021022.mm.1 NM_001009949.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021022.mm.1 TC0400002578.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021023.mm.1 NM_001009949.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021023.mm.1 NM_001009949.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021023.mm.1 TC0400002578.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021030.mm.1 NM_001009949.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021030.mm.1 NM_001009949.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021030.mm.1 TC0400002578.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 474720 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0