This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341023_m1
See other MCTP1 GE Assays ›
Gene Symbol:
Mctp1
Gene Name:
multiple C2 domains, transmembrane 1
Gene Aliases:
2810465F10Rik
Chromosome Location:
Chr.13: 76384961 - 77033284 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341023_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mctp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030174.2 NP_084450.2 3 - 4 554 85
GenBank mRNA AK047562.1 - 3 - 4 555 85
AK144765.1 - 3 - 4 353 85
AK154322.1 - 3 - 4 408 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000928.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mctp1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006023.mm.1 NM_030174.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006023.mm.1 NM_030174.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300006023.mm.1 TC1300000928.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0