This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341000_m1
See other FKBP9 GE Assays ›
Gene Symbol:
Fkbp9
Gene Name:
FK506 binding protein 9
Gene Aliases:
1810014L23, AW549641, FKBP-63, FKBP-9, FKBP60, FKBP63, FKBP65RS
Chromosome Location:
Chr.6: 56832059 - 56879360 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341000_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fkbp9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012056.2 NP_036186.2 4 - 5 843 90
GenBank mRNA AF090334.1 - 4 - 5 769 90
AK007499.1 - 2 - 3 233 90
AK189046.1 - 1 - 2 76 92
BC043129.1 - 4 - 5 843 6810029 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000678.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fkbp9

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
calcium ion binding
FK506 binding
isomerase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005260.mm.1 NM_012056.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005260.mm.1 NM_012056.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600005260.mm.1 TC0600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600005259.mm.1 NM_012056.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005259.mm.1 NM_012056.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600005259.mm.1 TC0600000678.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0