This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340929_m1
See other AI314180 GE Assays ›
Gene Symbol:
AI314180
Gene Name:
expressed sequence AI314180
Gene Aliases:
AW558785, BB181316, mKIAA0368
Chromosome Location:
Chr.4: 58797237 - 58913123 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340929_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AI314180
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172381.2 NP_759013.2 26 - 27 3209 62
GenBank mRNA AK035657.1 - 5 - 6 568 62
AK043020.1 - 27 - 28 3126 62
AK129127.1 - 9 - 10 1149 62
BC058684.1 - 26 - 27 3209 6834912 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002743.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AI314180

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

ER-associated ubiquitin-dependent protein catabolic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400022209.mm.1 NM_172381.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022209.mm.1 NM_172381.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400022209.mm.1 TC0400002743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400022208.mm.1 NM_172381.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022208.mm.1 NM_172381.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400022208.mm.1 TC0400002743.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0