This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01339718_m1
See other PPP1R13B GE Assays ›
Gene Symbol:
Ppp1r13b
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 13B
Gene Aliases:
AI449786, ASPP1, AW545810, Tp53bp2, Trp53bp2, p85
Chromosome Location:
Chr.12: 111828458 - 111908142 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01339718_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r13b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011625.1 NP_035755.1 15 - 16 3205 66
GenBank mRNA BC034207.1 - 11 - 12 2651 66
BC053092.1 - 15 - 16 3231 5719383 66
BC053732.1 - 11 - 12 2651 66
BC054788.1 - 15 - 16 3205 6826065 66
U58881.1 - 2 - 3 191 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002416.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r13b

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
negative regulation of cell cycle
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of neuron death
transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200016787.mm.1 NM_011625.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200016787.mm.1 NM_011625.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200016787.mm.1 TC1200002416.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0