This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337976_g1
See other RMDN3 GE Assays ›
Gene Symbol:
Rmdn3
Gene Name:
regulator of microtubule dynamics 3
Gene Aliases:
1200015F23Rik, AI131757, Fam82a2, Ptpip51, RMD-3, Rmd3
Chromosome Location:
Chr.2: 119136998 - 119157034 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337976_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rmdn3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033136.3 NP_001028308.1 10 - 11 1360 98
GenBank mRNA AK004786.1 - 10 - 11 1286 98
AK146299.1 - 10 - 11 1279 98
AK151090.1 - 10 - 11 1281 98
AK153322.1 - 10 - 11 1281 98
AK153369.1 - 10 - 11 1280 98
AK171770.1 - 10 - 11 1361 98
BC055754.1 - 10 - 11 1254 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004423.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
apoptotic process
cell differentiation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200036931.mm.1 NM_001033136.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200036931.mm.1 NM_001033136.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200036931.mm.1 TC0200004423.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0