This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337940_gH
See other PAK6 GE Assays ›
Gene Symbol:
Pak6
Gene Name:
p21 protein (Cdc42/Rac)-activated kinase 6
Gene Aliases:
4732456M09
Chromosome Location:
Chr.2: 118663027 - 118698020 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337940_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pak6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033254.3 NP_001028426.2 8 - 9 2262 86
NM_001145854.1 NP_001139326.1 7 - 8 1787 86
GenBank mRNA AK145601.1 - 7 - 8 1761 86
AK158913.1 - 8 - 9 2536 86
AK158944.1 - 8 - 9 2536 86
BC059912.1 - 8 - 9 2068 86
BC079572.1 - 8 - 9 2259 86
BC150754.1 - 8 - 9 2152 9087665 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001743.mm.1

Set Membership:

Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
protein phosphorylation
Rho protein signal transduction
regulation of mitotic cell cycle
learning
memory
locomotory behavior
phosphorylation
cell migration
signal transduction by protein phosphorylation
actin cytoskeleton organization
stress-activated protein kinase signaling cascade
activation of protein kinase activity
regulation of apoptotic process
regulation of MAPK cascade
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
Rac GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200012921.mm.1 NM_001033254.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012921.mm.1 NM_001033254.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200012921.mm.1 NM_001145854.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012921.mm.1 NM_001145854.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012921.mm.1 TC0200001743.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0