Assay Details
Gene Symbol: Prdx6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177256.5 | NP_796230.1 | 1 | 2764 | 150 | |
GenBank mRNA | AK077012.1 | - | 1 | 2760 | 150 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210753 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45486 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151509 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P187023 | AK077012.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483301 | BC087895.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483301 | AK161395.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483301 | NM_177256.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483301 | AK029613.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P483301 | AK077012.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21373887 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151509 | - | - |
Whole Mouse Genome G4122A | A_51_P483301 | BC087895.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483301 | AK161395.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483301 | NM_177256.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483301 | AK029613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P483301 | AK077012.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P187023 | NM_177256.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P187023 | AK077012.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138161 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139690 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151509 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02070741 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01491503 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02070738 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01959598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21373887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02101109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373849 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02068202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02101107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373843 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02207264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01719272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02008053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01891744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01891742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02070733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02070742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01892189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01891747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373851 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02207267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373845 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01279599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01809735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01279603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01279597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373927 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01681494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373841 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373829 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01279601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02209504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373858 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01681496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02209508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21373879 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473188 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473189 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447715_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437412_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473187 | - | - |
Mouse Genome 430 2.0 Array | 1437412_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447715_x_at | - | - |
Mouse Genome 430 2.0 Array | 1437412_at | - | - |
Mouse Genome 430 2.0 Array | 1447715_PM_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320769_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320769_PM_at | - | - |
Mouse Expression Set 430 | 1447715_PM_x_at | - | - |
Mouse Expression Set 430 | 1437412_PM_at | - | - |
Mouse Expression Set 430 | 1447715_x_at | - | - |
Mouse Expression Set 430 | 1437412_at | - | - |
Murine Genome U74v2 Set | 100332_PM_s_at | - | - |
Murine Genome U74v2 Set | 100332_s_at | - | - |
Murine Genome U74v2 Set | 166187_f_at | - | - |
Murine Genome U74v2 Set | 165881_f_at | - | - |
Murine Genome U74v2 Set | 166187_PM_f_at | - | - |
Murine Genome U74v2 Set | 165881_PM_f_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447715_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437412_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447715_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437412_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 516109 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2690193 | - | - |
MouseWG-6 v2 Expression BeadChip | 730315 | - | - |
MouseWG-6 v2 Expression BeadChip | 2810066 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 730315 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2760538 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2810066 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000730315 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002760538 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002810066 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733146 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244647 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239281 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2733146 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244647 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1239281 | - | - |
mouse-6 v1.1 (Illumina) | scl00320769.2_0-S | - | - |
mouse-6 v1.1 (Illumina) | scl00320769.2_294-S | - | - |
mouse-6 v1.1 (Illumina) | scl20713.1.955_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2690193 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2760538 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 730315 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2810066 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0320769.2_141-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20713.1.955_25-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00320769.2_0-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00320769.2_294-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810066 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 730315 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2690193 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760538 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244647 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2733146 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2797403 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1239281 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2797403 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244647 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239281 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733146 | - | - |
