This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337145_g1
See other KCNAB3 GE Assays ›
Gene Symbol:
Kcnab3
Gene Name:
potassium voltage-gated channel, shaker-related subfamily, beta member 3
Gene Aliases:
C330022D06Rik, Kcnab4, mKv(beta)4
Chromosome Location:
Chr.11: 69326258 - 69333041 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337145_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kcnab3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010599.4 NP_034729.3 3 - 4 749 65
GenBank mRNA AK029750.1 - 2 - 3 338 65
AK031806.1 - 3 - 4 730 65
AK044102.1 - 3 - 4 704 65
AK046194.1 - 3 - 4 739 65
AK049316.1 - 3 - 4 750 65
AK083134.1 - 3 - 4 751 65
BC017518.1 - 3 - 4 675 4507029 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000908.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kcnab3

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
regulation of ion transmembrane transport
voltage-gated ion channel activity
voltage-gated potassium channel activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100007688.mm.1 NM_010599.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007688.mm.1 NM_010599.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100007688.mm.1 TC1100000908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0