This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336563_m1
See other GRB14 GE Assays ›
Gene Symbol:
Grb14
Gene Name:
growth factor receptor bound protein 14
Gene Aliases:
AI505286
Chromosome Location:
Chr.2: 64912476 - 65022766 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336563_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grb14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016719.1 NP_057928.1 5 - 6 794 63
GenBank mRNA AF155647.1 - 5 - 6 794 63
AK010849.1 - 5 - 6 811 63
AK010903.1 - 5 - 6 811 63
AK147041.1 - 5 - 6 803 63
BC138581.1 - 5 - 6 700 8861601 63
BC145913.1 - 5 - 6 700 40131216 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003654.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grb14

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
insulin receptor signaling pathway
positive regulation of signal transduction
negative regulation of insulin receptor signaling pathway
SH3/SH2 adaptor activity
protein binding
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200031104.mm.1 NM_016719.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200031104.mm.1 NM_016719.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200031104.mm.1 TC0200003654.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0