This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336312_m1
See other UPF2 GE Assays ›
Gene Symbol:
Upf2
Gene Name:
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases:
-
Chromosome Location:
Chr.2: 5951447 - 6056703 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336312_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Upf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081132.1 NP_001074601.1 12 - 13 2462 71
GenBank mRNA BC039278.1 - 1 - 2 125 71
BC138358.1 - 12 - 13 2425 8861378 71
BC138359.1 - 12 - 13 2425 8861379 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
liver development
response to unfolded protein
organ regeneration

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0