This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336302_g1
See other SLC43A3 GE Assays ›
Gene Symbol:
Slc43a3
Gene Name:
solute carrier family 43, member 3
Gene Aliases:
Eeg1, SEEEG-1
Chromosome Location:
Chr.2: 84936545 - 84958509 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336302_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc43a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021398.3 NP_067373.2 2 - 3 549 91
GenBank mRNA AF188622.1 - 2 - 3 543 91
AK086962.1 - 1 - 2 978 91
BC089603.1 - 1 - 2 249 6758493 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001226.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc43a3

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s)

transmembrane transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200009804.mm.1 NM_021398.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009804.mm.1 NM_021398.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200009804.mm.1 TC0200001226.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200009805.mm.1 NM_021398.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009805.mm.1 NM_021398.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200009805.mm.1 TC0200001226.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0