This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335894_m1
See other ZIM3 GE Assays ›
Gene Symbol:
Zim3
Gene Name:
zinc finger, imprinted 3
Gene Aliases:
1700128I23Rik
Chromosome Location:
Chr.7: 6955685 - 6976662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335894_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zim3
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036631.2
4 - 5 433 71
GenBank mRNA AF365932.1 - LIT1822
Zim3
4 - 5 438 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 3099 Mir3099 mir-3099,
mmu-mir-3099
-162.3kb Anti-sense Chr. 7 - 6803587 - 6803670 on NCBI Build GRCm38
Downstream zinc finger protein 264 Zfp264 4.1kb Anti-sense Chr. 7 - 6977413 - 6985640 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002219.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700020152.mm.1 NR_036631.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700020152.mm.1 NR_036631.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700020152.mm.1 TC0700002219.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0