This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335464_m1
See other 4930559C10RIK GE Assays ›
Gene Symbol:
4930559C10Rik
Gene Name:
RIKEN cDNA 4930559C10 gene
Gene Aliases:
-
Chromosome Location:
Chr.12: 93416457 - 93444823 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335464_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930559C10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016182.1 - 2 - 3 247 94
BY715789.1 - 2 - 3 242 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sel-1 suppressor of lin-12-like (C. elegans) Sel1l AW493766,
Sel1h,
mKIAA4137
-1.6mb Sense Chr. 12 - 91806043 - 91849157 on NCBI Build GRCm38
Downstream microRNA 8099-2 Mir8099-2 Gm27692,
mmu-mir-8099-2
496.9kb Anti-sense Chr. 12 - 93927338 - 93927464 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514470 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579701 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371767 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490240 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264480 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836692 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803384 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956741 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8375 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07710181 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133040 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644921 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644934 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644910 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644926 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644919 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10197142 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644927 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644920 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644932 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644908 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10197140 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644915 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644916 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644931 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644923 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28644924 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0