This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01334908_m1
See other SERPINA10 GE Assays ›
Gene Symbol:
Serpina10
Gene Name:
serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
Gene Aliases:
PZI, ZPI
Chromosome Location:
Chr.12: 103616675 - 103631490 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01334908_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serpina10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301404.1 NP_001288333.1 1 - 2 60 85
NM_144834.4 NP_659083.2 1 - 2 60 85
GenBank mRNA AK050552.1 - 34 85
AY324633.1 - 1 - 2 60 85
BC018416.1 - 1 - 2 37 4196269 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY324633.1, BC018416.1, NM_001301404.1, NM_144834.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serpina10

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
hemostasis
negative regulation of peptidase activity
negative regulation of endopeptidase activity
serine-type endopeptidase inhibitor activity
heparin binding
peptidase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200015775.mm.1 NM_144834.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200015775.mm.1 TC1200002271.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0