This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01334598_m1
See other CTNND1 GE Assays ›
Gene Symbol:
Ctnnd1
Gene Name:
catenin (cadherin associated protein), delta 1
Gene Aliases:
AA409437, AU019353, Catns, Ctnnd, P120, mKIAA0384
Chromosome Location:
Chr.2: 84600781 - 84658972 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01334598_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctnnd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085448.1 NP_001078917.1 5 - 6 1388 75
NM_001085449.1 NP_001078918.1 4 - 5 1099 75
NM_001085450.1 NP_001078919.1 5 - 6 1388 75
NM_001085453.1 NP_001078922.1 5 - 6 1158 75
NM_007615.4 NP_031641.2 6 - 7 1506 75
GenBank mRNA AB093237.1 - 6 - 7 1222 75
AK133193.1 - 5 - 6 1109 75
AK148871.1 - 1 - 2 169 75
AK161790.1 - 6 - 7 1282 75
AK164485.1 - 5 - 6 1373 75
AK168453.1 - 4 - 5 1097 75
BC043108.1 - 5 - 6 1323 6406902 75
BC046589.1 - 4 - 5 899 75
BC054544.1 - 6 - 7 1439 6408956 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003869.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctnnd1

Panther Classification:

Molecular Function -

intermediate filament binding protein

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of a polarized epithelium
transcription, DNA-templated
regulation of transcription, DNA-templated
cell adhesion
salivary gland morphogenesis
Wnt signaling pathway
single organismal cell-cell adhesion
keratinocyte differentiation
epithelial cell differentiation involved in salivary gland development
negative regulation of canonical Wnt signaling pathway
receptor binding
protein binding
protein kinase binding
protein phosphatase binding
protein domain specific binding
cadherin binding
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085448.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085448.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085449.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085449.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085450.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085450.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_001085453.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_007615.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 NM_007615.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200033465.mm.1 TC0200003869.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm