This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333207_m1
See other MCM10 GE Assays ›
Gene Symbol:
Mcm10
Gene Name:
minichromosome maintenance deficient 10 (S. cerevisiae)
Gene Aliases:
2410041F14Rik, AU018508, C330019M07Rik, C79164
Chromosome Location:
Chr.2: 4990724 - 5012848 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333207_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mcm10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027290.3 NP_081566.2 8 - 9 1311 92
GenBank mRNA AK030780.1 - 8 - 9 1256 92
AK041406.1 - 8 - 9 1246 92
AK041749.1 - 2 - 3 317 92
BC064314.1 - 8 - 9 1674 92
BC094576.1 - 8 - 9 1311 6853824 92
BC120687.1 - 7 - 8 1220 40129610 92
BC120689.1 - 7 - 8 1220 40129612 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002869.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mcm10

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA replication initiation
cell proliferation
DNA binding
DNA replication origin binding
double-stranded DNA binding
single-stranded DNA binding
identical protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200023802.mm.1 NM_027290.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023802.mm.1 NM_027290.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200023802.mm.1 TC0200002869.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200023800.mm.1 NM_027290.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023800.mm.1 NM_027290.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200023800.mm.1 TC0200002869.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0