This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333023_m1
See other 5730457N03RIK GE Assays ›
Gene Symbol:
5730457N03Rik
Gene Name:
RIKEN cDNA 5730457N03 gene
Gene Aliases:
Evx1as
Chromosome Location:
Chr.6: 52308389 - 52314832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333023_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5730457N03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038163.1 1 - 2 67 82
GenBank mRNA AK031498.1 - 1 - 2 69 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB620958.1 - 1 - 2 69 Chr. 6 - 52310648 - 52314832 [-] on Build GRCm38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Hoxa distal transcript antisense RNA Hottip ENSMUSG00000055408,
Gm15053,
OTTMUSG00000018874
-45.7kb Anti-sense Chr. 6 - 52262775 - 52267603 on NCBI Build GRCm38
Downstream RIKEN cDNA 1700094M24 gene 1700094M24Rik 177.6kb Anti-sense Chr. 6 - 52492451 - 52500085 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002288.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
chromatin binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600017942.mm.1 NR_038163.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017942.mm.1 NR_038163.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017942.mm.1 TC0600002288.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017943.mm.1 NR_038163.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017943.mm.1 NR_038163.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017943.mm.1 TC0600002288.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017941.mm.1 NR_038163.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017941.mm.1 NR_038163.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017941.mm.1 TC0600002288.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0