This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01332211_m1
See other ADARB2 GE Assays ›
Gene Symbol:
Adarb2
Gene Name:
adenosine deaminase, RNA-specific, B2
Gene Aliases:
Adar3, RED2
Chromosome Location:
Chr.13: 8202318 - 8768762 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01332211_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adarb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289530.1 NP_001276459.1 5 - 6 1732 106
NM_052977.5 NP_443209.2 5 - 6 1732 106
NR_110345.1 5 - 6 1732 106
GenBank mRNA AF232942.1 - 5 - 6 1349 106
AF270495.1 - 5 - 6 1449 106
AK033916.1 - 5 - 6 1671 106
AK045538.1 - 5 - 6 1671 106
AK046083.1 - 5 - 6 1676 106
AK134964.1 - 5 - 6 1734 106
AK139079.1 - 5 - 6 1690 106
AK139273.1 - 5 - 6 1690 106
BC052426.1 - 5 - 6 1657 6835942 106
BC059822.1 - 5 - 6 1685 6844585 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000037.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adarb2

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
RNA binding
adenosine deaminase activity
hydrolase activity
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300000278.mm.1 NM_001289530.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000278.mm.1 NM_052977.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300000278.mm.1 NR_110345.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000278.mm.1 TC1300000037.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0