This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01331054_m1
See other ACVR1C GE Assays ›
Gene Symbol:
Acvr1c
Gene Name:
activin A receptor, type IC
Gene Aliases:
ACVRLK7, ALK7, Alk-7, C230097P10
Chromosome Location:
Chr.2: 58267453 - 58357896 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01331054_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acvr1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033369.3 NP_001028541.1 4 - 5 655 107
NM_001111030.1 NP_001104500.1 5 - 6 1022 107
GenBank mRNA AK049089.1 - 4 - 5 656 107
AK142396.1 - 5 - 6 1026 107
BC028780.1 - 1 - 2 40 107
BC145224.1 - 4 - 5 818 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003576.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
trophectodermal cell proliferation
response to dietary excess
protein phosphorylation
apoptotic process
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor complex assembly
determination of left/right symmetry
response to glucose
phosphorylation
lipid storage
cell differentiation
apoptotic nuclear changes
response to insulin
activin receptor signaling pathway
nodal signaling pathway
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of insulin secretion
negative regulation of trophoblast cell migration
negative regulation of chorionic trophoblast cell proliferation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
kinase activity
activin receptor activity, type I
transferase activity
transferase activity, transferring phosphorus-containing groups
growth factor binding
nodal binding
SMAD binding
metal ion binding
activin binding
BMP receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200030450.mm.1 NM_001033369.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030450.mm.1 NM_001033369.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200030450.mm.1 NM_001111030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030450.mm.1 NM_001111030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030450.mm.1 TC0200003576.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0