This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327386_m1
See other GPR155 GE Assays ›
Gene Symbol:
Gpr155
Gene Name:
G protein-coupled receptor 155
Gene Aliases:
1110017O10Rik, DEPDC3, F730029F15Rik, PGR22
Chromosome Location:
Chr.2: 73341506 - 73386851 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327386_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr155
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190297.2 NP_001177226.1 1 - 2 217 102
NM_001276443.1 NP_001263372.1 1 - 2 217 102
NM_001276444.1 NP_001263373.1 1 - 2 217 102
GenBank mRNA AK089436.1 - 1 - 2 135 102
BC115698.1 - 1 - 2 113 40060105 102
HM161763.2 - 1 - 2 194 102
HM161764.2 - 1 - 2 194 102
HM161765.2 - 1 - 2 194 102
HM161766.2 - 1 - 2 194 102
KF358283.1 - 1 - 2 194 102
KF358284.1 - 1 - 2 194 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC115698.1, HM161763.2, HM161764.2, HM161765.2, HM161766.2, KF358283.1, KF358284.1, NM_001190297.2, NM_001276443.1, NM_001276444.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

cognition
molecular_function

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 324508 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0