This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327381_g1
See other GPR155 GE Assays ›
Gene Symbol:
Gpr155
Gene Name:
G protein-coupled receptor 155
Gene Aliases:
1110017O10Rik, DEPDC3, F730029F15Rik, PGR22
Chromosome Location:
Chr.2: 73341506 - 73386851 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327381_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr155
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190297.2 NP_001177226.1 12 - 13 2283 56
NM_001276443.1 NP_001263372.1 12 - 13 2283 56
NM_001276444.1 NP_001263373.1 11 - 12 2199 56
GenBank mRNA AK089436.1 - 12 - 13 2201 56
AY255560.1 - 3 - 4 399 56
BC026382.1 - 2 - 3 299 56
BC115698.1 - 12 - 13 2179 40060105 56
HM161763.2 - 12 - 13 2260 56
HM161764.2 - 12 - 13 2260 56
HM161765.2 - 12 - 13 2260 56
HM161766.2 - 11 - 12 2176 56
KF358283.1 - 12 - 13 2260 56
KF358284.1 - 11 - 12 2142 56

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

cognition
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0