This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01322956_m1
See other LGALS3 GE Assays ›
Gene Symbol:
Lgals3
Gene Name:
lectin, galactose binding, soluble 3
Gene Aliases:
GBP, L-34, Mac-2, gal3
Chromosome Location:
Chr.14: 47373860 - 47386167 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01322956_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lgals3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145953.1 NP_001139425.1 2 - 3 138 67
GenBank mRNA AK149783.1 - 2 - 3 85 67
AK159536.1 - 2 - 3 79 67
BC132328.1 - 2 - 3 64 40130605 67
BC138790.1 - 2 - 3 64 8861812 67
BC145419.1 - 2 - 3 64 9053977 67
X16834.1 - 2 - 3 57 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK149783.1, AK159536.1, BC132328.1, BC138790.1, BC145419.1, NM_001145953.1, X16834.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
immune system process
monocyte chemotaxis
mRNA processing
signal transduction
positive regulation of cell proliferation
RNA splicing
positive regulation of serotonin secretion
cell differentiation
extracellular matrix organization
neutrophil chemotaxis
regulation of T cell proliferation
negative regulation of apoptotic process
innate immune response
positive regulation of angiogenesis
negative regulation of endocytosis
eosinophil chemotaxis
macrophage chemotaxis
negative regulation of T cell receptor signaling pathway
positive chemotaxis
regulation of T cell apoptotic process
mononuclear cell migration
positive regulation of mononuclear cell migration
positive regulation of calcium ion import
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of cell proliferation in bone marrow
negative regulation of immunological synapse formation
negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
negative regulation of extrinsic apoptotic signaling pathway
protein binding
IgE binding
carbohydrate binding
Fc-gamma receptor I complex binding
chemoattractant activity
laminin binding
poly(A) RNA binding
monosaccharide binding
disaccharide binding
advanced glycation end-product receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0