This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01321710_m1
See other CNIH1 GE Assays ›
Gene Symbol:
Cnih1
Gene Name:
cornichon homolog 1 (Drosophila)
Gene Aliases:
0610007J15, CNIH-1, Cnih
Chromosome Location:
Chr.14: 46775567 - 46788888 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01321710_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnih1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009919.2 NP_034049.2 3 - 4 353 69
GenBank mRNA AK002293.1 - 3 - 4 321 69
AK011200.1 - 3 - 4 306 69
AK152432.1 - 3 - 4 304 69
AK154479.1 - 3 - 4 304 69
AK168154.1 - 3 - 4 304 69
BC034868.1 - 3 - 4 313 2938052 69
DV653213.1 - 3 - 4 407 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001920.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cnih1

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
vesicle-mediated transport
intracellular signal transduction
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400014504.mm.1 NM_009919.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400014504.mm.1 NM_009919.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400014504.mm.1 TC1400001920.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header