This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319962_m1
See other AK5 GE Assays ›
Gene Symbol:
Ak5
Gene Name:
adenylate kinase 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 152462815 - 152668376 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319962_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ak5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081277.1 NP_001074746.1 5 - 6 980 69
GenBank mRNA AK053807.1 - 5 - 6 980 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003147.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
phosphorylation
ATP metabolic process
nucleotide binding
adenylate kinase activity
nucleoside diphosphate kinase activity
ATP binding
kinase activity
transferase activity
nucleobase-containing compound kinase activity
nucleoside kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024705.mm.1 NM_001081277.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024705.mm.1 NM_001081277.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024705.mm.1 TC0300003147.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0