This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318965_m1
See other MMP14 GE Assays ›
Gene Symbol:
Mmp14
Gene Name:
matrix metallopeptidase 14 (membrane-inserted)
Gene Aliases:
AI325305, MT-MMP-1, MT1-MMP, sabe
Chromosome Location:
Chr.14: 54431604 - 54441258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318965_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmp14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008608.3 NP_032634.3 4 - 5 951 110
GenBank mRNA AK040968.1 - 924 110
AK088476.1 - 4 - 5 951 110
AK134340.1 - 4 - 5 882 110
AK138611.1 - 4 - 5 892 110
AK149636.1 - 4 - 5 880 110
AK149907.1 - 4 - 5 931 110
AK149988.1 - 4 - 5 851 110
AK155117.1 - 4 - 5 953 110
AK163601.1 - 4 - 5 943 110
AK165014.1 - 4 - 5 952 110
AK180055.1 - 1 - 2 187 110
AK198345.1 - 1 - 2 188 110
AK209696.1 - 1 - 2 90 110
BC076638.1 - 4 - 5 943 30658833 110
DQ249870.1 - 4 - 5 689 110
U54984.1 - 4 - 5 935 110
X83536.2 - 4 - 5 861 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000745.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mmp14

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

ossification
endochondral ossification
regulation of transcription, DNA-templated
proteolysis
response to hormone
positive regulation of peptidase activity
cell migration
positive regulation of cell growth
lung development
positive regulation of cell migration
collagen catabolic process
zymogen activation
chondrocyte proliferation
embryonic cranial skeleton morphogenesis
branching morphogenesis of an epithelial tube
tissue remodeling
negative regulation of focal adhesion assembly
bone development
craniofacial suture morphogenesis
transcription factor activity, sequence-specific DNA binding
metalloendopeptidase activity
integrin binding
calcium ion binding
peptidase activity
metallopeptidase activity
zinc ion binding
peptidase activator activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400005814.mm.1 NM_008608.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400005814.mm.1 TC1400000745.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0