This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312039_m1
See other PDE10A GE Assays ›
Gene Symbol:
Pde10a
Gene Name:
phosphodiesterase 10A
Gene Aliases:
-
Chromosome Location:
Chr.17: 8525187 - 8986648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312039_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde10a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290707.1 NP_001277636.1 13 - 14 1470 85
NM_011866.2 NP_035996.2 12 - 13 1424 85
GenBank mRNA AF110507.1 - 12 - 13 1295 85
AK039249.1 - 13 - 14 1470 85
AK147690.1 - 11 - 12 1341 85
AK162221.1 - 12 - 13 1079 85
AK162804.1 - 10 - 11 965 85
AY360383.1 - 12 - 13 1424 85
BC113201.1 - 12 - 13 1290 40048238 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde10a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
metabolic process
regulation of protein kinase A signaling
regulation of cAMP-mediated signaling
cGMP catabolic process
nucleotide binding
catalytic activity
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
drug binding
hydrolase activity
cAMP binding
cGMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0